Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5351 |
Gene Name: | procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 |
Gene Aliases: |
EDS6, LH, LH1, LLH, PLOD |
Location: |
Chr.1:11934667-11975542 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLOD1 | NM_000302.3 | 4 | 490 | NP_000293.2 |
NM_001316320.1 | 5 | 631 | NP_001303249.1 | |
AK294688.1 | ||||
AK295254.1 | 4 | 434 | ||
AK299150.1 | 5 | 564 | ||
AK312243.1 | 4 | 423 | ||
AK316285.1 | 5 | 567 | ||
AU139472.1 | 4 | 469 | ||
BC016657.2 | 4 | 394 | AAH16657.1 | |
DC334246.1 | 4 | 490 | ||
M98252.1 | 4 | 401 |
Set Membership: |
Intragenic Exonic non-DGV Variation |