Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8929 |
Gene Name: | paired like homeobox 2b |
Gene Aliases: |
NBLST2, NBPhox, PMX2B |
Location: |
Chr.4:41744082-41748970 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHOX2B | NM_003924.3 | 3 | 1434 | NP_003915.2 |
BC017199.2 | 3 | 1170 | AAH17199.1 | |
D82344.1 | 3 | 1434 | BAA11555.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32898 | Chr.4:41744516 - 41746501 on Build GRCh38 | Gain | PHOX2B |
Set Membership: |
Intragenic Exonic DGV Variation |