Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 7274 |
Gene Name: | alpha tocopherol transfer protein |
Gene Aliases: |
ATTP, AVED, TTP1, alphaTTP |
Location: |
Chr.8:63058409-63086523 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTPA | NM_000370.3 | 5 | 1128 | NP_000361.1 |
XM_006716468.3 | 3 | 1250 | XP_006716531.1 | |
BC058000.1 | 5 | 1132 | AAH58000.1 | |
D49488.1 | 5 | 1114 | BAA08449.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv611448 | Chr.8:63038678 - 63167927 on Build GRCh38 | Loss | YTHDF3-AS1 GGH TTPA |
Set Membership: |
Intragenic Exonic DGV Variation |