Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 351 |
Gene Name: | amyloid beta precursor protein |
Gene Aliases: |
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2 |
Location: |
Chr.21:25880550-26171128 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APP | NM_000484.3 | 14 | 1982 | NP_000475.1 |
NM_001136016.3 | 13 | 1906 | NP_001129488.1 | |
NM_001136129.2 | 11 | 1589 | NP_001129601.1 | |
NM_001136130.2 | 13 | 1814 | NP_001129602.1 | |
NM_001136131.2 | 12 | 1644 | NP_001129603.1 | |
NM_001204301.1 | 14 | 1982 | NP_001191230.1 | |
NM_001204302.1 | 13 | 1925 | NP_001191231.1 | |
NM_001204303.1 | 12 | 1757 | NP_001191232.1 | |
NM_201413.2 | 13 | 1925 | NP_958816.1 | |
NM_201414.2 | 12 | 1757 | NP_958817.1 | |
A02759.1 | 12 | 1703 | CAA00268.1 | |
AF282245.1 | 11 | 1516 | AAQ14327.1 | |
AK294534.1 | 13 | 1761 | ||
AK295373.1 | 4 | 482 | ||
AK295621.1 | 13 | 1867 | ||
AK296229.1 | 12 | 1644 | ||
AK297229.1 | 10 | 1120 | ||
AK297412.1 | 11 | 1486 | ||
AK298861.1 | 11 | 1659 | ||
AK312326.1 | 14 | 1847 | ||
BC065529.1 | 13 | 1856 | AAH65529.1 | |
M16765.1 | 7 | 717 | AAA51722.1 | |
M18734.1 | 3 | 265 | AAA51726.1 | |
X06989.1 | 13 | 1849 | CAA30050.1 | |
Y00264.1 | 12 | 1704 | CAA68374.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |