Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2189 |
Gene Name: | Fanconi anemia complementation group G |
Gene Aliases: |
FAG, XRCC9 |
Location: |
Chr.9:35073838-35080016 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FANCG | NM_004629.1 | 14 | 2470 | NP_004620.1 |
AJ007669.1 | 14 | 2470 | CAA07602.1 | |
AK293427.1 | ||||
AK311348.1 | 13 | 1944 | ||
BC000032.2 | 14 | 2116 | AAH00032.1 | |
BC011623.2 | 14 | 2243 | AAH11623.1 | |
KU159422.1 | 14 | 2483 | ||
U70310.1 | 14 | 2112 | AAB80802.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |