Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5429 |
Gene Name: | DNA polymerase eta |
Gene Aliases: |
RAD30, RAD30A, XP-V, XPV |
Location: |
Chr.6:43576141-43620523 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLH | NM_001291969.1 | 8 | 1104 | NP_001278898.1 |
NM_001291970.1 | 10 | 1463 | NP_001278899.1 | |
NM_006502.2 | 10 | 1463 | NP_006493.1 | |
XM_005249186.3 | 9 | 1316 | XP_005249243.1 | |
XM_011514698.2 | 9 | 1237 | XP_011513000.1 | |
AB024313.1 | 10 | 1396 | BAA81666.1 | |
AF158185.1 | 10 | 1324 | AAD43810.1 | |
AJ420484.1 | 7 | 780 | ||
AK023893.1 | 10 | 1454 | ||
AK122807.1 | 7 | 1371 | ||
AK294430.1 | 10 | 1444 | ||
AK307660.1 | 8 | 1089 | ||
AK316151.1 | 9 | 1278 | ||
BC015742.1 | 10 | 1307 | AAH15742.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |