Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55063 |
Gene Name: | zinc finger CW-type and PWWP domain containing 1 |
Gene Aliases: |
ZCW1 |
Location: |
Chr.7:100400826-100429239 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZCWPW1 | NM_001258008.1 | NP_001244937.1 | ||
NM_017984.4 | NP_060454.3 | |||
XM_005250479.2 | XP_005250536.1 | |||
XM_005250480.2 | XP_005250537.1 | |||
XM_006716035.3 | XP_006716098.1 | |||
XM_006716036.2 | XP_006716099.1 | |||
XM_006716037.3 | XP_006716100.1 | |||
XM_006716038.3 | XP_006716101.1 | |||
XM_006716040.3 | XP_006716103.1 | |||
XM_017012379.1 | XP_016867868.1 | |||
XM_017012380.1 | XP_016867869.1 | |||
AK000919.1 | BAA91424.1 | |||
AK093038.1 | BAC04028.1 | |||
AK300745.1 | ||||
AK302108.1 | ||||
AL136735.1 | CAB66669.1 | |||
BC002725.1 | AAH02725.1 | |||
KU178677.1 | ||||
KU178678.1 | ||||
KU178679.1 | ||||
KU178680.1 | ||||
KU178681.1 | ||||
KU178682.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | EPHB4 AP1S1 PILRB CLDN15 AGFG2 ZNHIT1 PCOLCE-AS1 NAT16 STAG3L5P MUC17 MIR6875 MIR6840 SERPINE1 MUC3A ACTL6B ZAN ZCWPW1 GATS MOGAT3 SAP25 NYAP1 AZGP1P1 C7orf61 FBXO24 SLC12A9 GPC2 TFR2 PVRIG2P MCM7 ZKSCAN1 TSC22D4 PPP1R35 ZNF3 PLOD3 ACHE C7orf43 STAG3 LOC102724094 MIR106B GAL3ST4 CNPY4 AZGP1 MEPCE MIR25 EPO ZSCAN21 COPS6 POP7 MUC12 MIR4653 STAG3L5P-PVRIG2P-PILRB UFSP1 LAMTOR4 AP4M1 LOC105375429 SPDYE3 PMS2P1 MIR93 MBLAC1 PCOLCE TRIP6 MIR4658 MOSPD3 PVRIG VGF GIGYF1 GNB2 SRRT TAF6 TRIM56 LRCH4 ZASP PILRA FIS1 |
nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss | PMS2P1 ZCWPW1 NYAP1 PILRB C7orf61 MEPCE PVRIG2P STAG3L5P MIR6840 STAG3L5P-PVRIG2P-PILRB TSC22D4 PPP1R35 PILRA SPDYE3 |
Set Membership: |
Intragenic Non-exonic DGV Variation |