Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3028 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 10 |
Gene Aliases: |
17b-HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1 |
Location: |
Chr.X:53431258-53434376 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSD17B10 | NM_001037811.2 | NP_001032900.1 | ||
NM_004493.2 | NP_004484.1 | |||
AF035555.2 | AAC15902.1 | |||
AF069134.1 | AAC39900.1 | |||
AU132847.1 | ||||
BC000372.2 | AAH00372.1 | |||
BC008708.1 | AAH08708.1 | |||
CR456723.1 | CAG33004.1 | |||
KU178107.1 | ||||
KU178108.1 | ||||
U73514.1 | AAB68958.1 | |||
U96132.1 | AAC51812.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | MTRNR2L10 PAGE5 TSPYL2 SMC1A MIR6895 GNL3L RIBC1 FOXR2 HSD17B10 FGD1 FAM104B MIRLET7F2 MAGED2 LOC100421746 ITIH6 TSR2 KANTR SNORA109 IQSEC2 MIR6857 XAGE3 MIR4536-1 PFKFB1 ALAS2 HUWE1 MIR6894 MAGEH1 TRO GPR173 SNORA11 PAGE2 FAM156A FAM120C FAM156B WNK3 MIR4536-2 KDM5C USP51 PAGE2B APEX2 PHF8 PAGE3 MIR98 |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | MTRNR2L10 PAGE5 TSPYL2 SMC1A MIR6895 GNL3L RIBC1 FOXR2 HSD17B10 FGD1 FAM104B MIRLET7F2 MAGED2 LOC100421746 ITIH6 TSR2 KANTR SNORA109 IQSEC2 MIR6857 XAGE3 MIR4536-1 PFKFB1 ALAS2 HUWE1 MIR6894 MAGEH1 TRO GPR173 SNORA11 PAGE2 FAM156A FAM120C FAM156B WNK3 MIR4536-2 KDM5C USP51 PAGE2B APEX2 PHF8 PAGE3 MIR98 |
nsv6908 | Chr.X:53412544 - 53446505 on Build GRCh38 | Insertion | SMC1A RIBC1 HSD17B10 |
Set Membership: |
Intragenic Non-exonic DGV Variation |