Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2878 |
Gene Name: | glutathione peroxidase 3 |
Gene Aliases: |
GPx-P, GSHPx-3, GSHPx-P |
Location: |
Chr.5:151020438-151028993 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPX3 | NM_002084.3 | 5 | 828 | NP_002075.2 |
AF217787.1 | 5 | 615 | AAF43005.1 | |
AK310242.1 | 5 | 680 | ||
BC013601.1 | 5 | 658 | AAH13601.1 | |
BC035841.1 | 5 | 663 | AAH35841.1 | |
BC050378.1 | 5 | 663 | AAH50378.2 | |
BC066949.1 | 5 | 677 | AAH66949.1 | |
BI522602.1 | ||||
D00632.1 | 5 | 664 | BAA00525.1 | |
X58295.1 | 5 | 659 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526224 | Chr.5:151003841 - 151070675 on Build GRCh38 | Gain | GPX3 TNIP1 |
Set Membership: |
Intragenic Exonic DGV Variation |