Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5189 |
Gene Name: | peroxisomal biogenesis factor 1 |
Gene Aliases: |
HMLR1, PBD1A, PBD1B, ZWS, ZWS1 |
Location: |
Chr.7:92487023-92528531 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PEX1 | NM_000466.2 | 5 | 1183 | NP_000457.1 |
NM_001282677.1 | 5 | 1183 | NP_001269606.1 | |
NM_001282678.1 | 5 | 1218 | NP_001269607.1 | |
XM_017012319.1 | 1 | 186 | XP_016867808.1 | |
AB008112.1 | 5 | 1154 | BAA85162.1 | |
AB052090.1 | 5 | 1087 | BAB59061.1 | |
AB052091.1 | 5 | 1087 | BAB59062.1 | |
AB052092.1 | 5 | 1087 | BAB59063.1 | |
AB052093.1 | 5 | 1087 | BAB59064.1 | |
AB052094.1 | 5 | 1087 | BAB59065.1 | |
AF026086.1 | 5 | 1147 | AAB87880.1 | |
AF030356.1 | 5 | 1091 | AAB99758.1 | |
AK292955.1 | 5 | 1088 | ||
AK293757.1 | 5 | 1126 | ||
AK293848.1 | ||||
AK295686.1 | ||||
BC035575.1 | 5 | 1125 | AAH35575.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2657169 | Chr.7:92517344 - 92518793 on Build GRCh38 | Deletion | PEX1 |
esv3614141 | Chr.7:92517321 - 92518849 on Build GRCh38 | Loss | PEX1 |
Set Membership: |
Intragenic Exonic DGV Variation |