Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 1311 |
Gene Name: | cartilage oligomeric matrix protein |
Gene Aliases: |
EDM1, EPD1, MED, PSACH, THBS5, TSP5 |
Location: |
Chr.19:18782773-18791305 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COMP | NM_000095.2 | NP_000086.2 | ||
AB086984.1 | BAC53888.1 | |||
AK074508.1 | BAC11031.1 | |||
AK223216.1 | BAD96936.1 | |||
AK290595.1 | ||||
AK296586.1 | ||||
AK297798.1 | ||||
AK297814.1 | ||||
AU120397.1 | ||||
BC110847.1 | AAI10848.1 | |||
BC125092.1 | ||||
L32137.1 | AAA57253.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3643845 | Chr.19:18786208 - 18791888 on Build GRCh38 | Loss | COMP |
nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion | SUGP1 TM6SF2 COPE SLC25A42 HOMER3 BORCS8 NR2C2AP TMEM161A ARMC6 RFXANK BORCS8-MEF2B GDF1 CRTC1 SUGP2 HAPLN4 COMP CERS1 KLHL26 NCAN UPF1 DDX49 LOC102724360 MEF2B |
nsv952422 | Chr.19:18606691 - 18807891 on Build GRCh38 | Deletion | COMP KLHL26 TMEM59L CRTC1 CRLF1 |
nsv833777 | Chr.19:18765126 - 18920267 on Build GRCh38 | Loss | COPE COMP CERS1 UPF1 DDX49 GDF1 CRTC1 |
Set Membership: |
Intragenic Non-exonic DGV Variation |