Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5429 |
Gene Name: | DNA polymerase eta |
Gene Aliases: |
RAD30, RAD30A, XP-V, XPV |
Location: |
Chr.6:43576141-43620523 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLH | NM_001291969.1 | 3 | 528 | NP_001278898.1 |
NM_001291970.1 | 5 | 887 | NP_001278899.1 | |
NM_006502.2 | 5 | 887 | NP_006493.1 | |
XM_005249186.3 | 4 | 740 | XP_005249243.1 | |
XM_011514698.2 | 4 | 661 | XP_011513000.1 | |
AB024313.1 | 5 | 820 | BAA81666.1 | |
AF158185.1 | 5 | 748 | AAD43810.1 | |
AJ420484.1 | 2 | 204 | ||
AK023893.1 | 5 | 878 | ||
AK122807.1 | 2 | 795 | ||
AK294430.1 | 5 | 868 | ||
AK307660.1 | 3 | 513 | ||
AK316151.1 | 4 | 702 | ||
BC015742.1 | 5 | 731 | AAH15742.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |