Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8890 |
Gene Name: | eukaryotic translation initiation factor 2B subunit delta |
Gene Aliases: |
EIF-2B, EIF2B, EIF2Bdelta |
Location: |
Chr.2:27364352-27370457 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF2B4 | NM_001034116.1 | NP_001029288.1 | ||
NM_001318965.1 | NP_001305894.1 | |||
NM_001318966.1 | NP_001305895.1 | |||
NM_001318967.1 | NP_001305896.1 | |||
NM_001318968.1 | NP_001305897.1 | |||
NM_001318969.1 | NP_001305898.1 | |||
NM_015636.3 | NP_056451.3 | |||
NM_172195.3 | NP_751945.2 | |||
XM_006712132.1 | XP_006712195.1 | |||
XM_011533147.2 | XP_011531449.1 | |||
AB209532.1 | 3 | 1190 | BAD92769.1 | |
AF057699.1 | AAQ13438.1 | |||
AF112207.1 | AAF17195.1 | |||
AF218009.1 | AAG17251.1 | |||
AJ011305.1 | CAB57260.1 | |||
AJ011306.1 | CAB57261.1 | |||
AK300734.1 | ||||
AL050109.1 | CAB43277.1 | |||
BC001870.1 | AAH01870.1 | |||
BC091502.1 | AAH91502.1 | |||
BG717490.1 | ||||
BI255289.1 | ||||
JQ991007.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain | NRBP1 PPM1G TRIM54 FTH1P3 ZNF513 KRTCAP3 SLC30A3 LOC105374363 IFT172 MPV17 UCN GTF3C2-AS1 EIF2B4 GTF3C2 DNAJC5G SNX17 |
esv3575209 | Chr.2:27334578 - 27389381 on Build GRCh38 | Gain | PPM1G LOC105374363 ZNF513 GTF3C2-AS1 EIF2B4 GTF3C2 SNX17 |
nsv581244 | Chr.2:27335385 - 27480628 on Build GRCh38 | Loss | NRBP1 KRTCAP3 PPM1G FTH1P3 LOC105374363 IFT172 ZNF513 GTF3C2-AS1 EIF2B4 GTF3C2 SNX17 |
Set Membership: |
Intragenic Non-exonic DGV Variation |