Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4089 |
Gene Name: | SMAD family member 4 |
Gene Aliases: |
DPC4, JIP, MADH4, MYHRS |
Location: |
Chr.18:51030213-51085042 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMAD4 | NM_005359.5 | 6 | 1261 | NP_005350.1 |
AK290770.1 | 6 | 923 | ||
BC002379.2 | 7 | 956 | AAH02379.1 | |
BX647129.1 | 2 | 2725 | ||
KU178168.1 | 5 | 723 | ||
KU178169.1 | 5 | 723 | ||
U44378.1 | 5 | 851 | AAA91041.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |