Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 146059 |
Gene Name: | codanin 1 |
Gene Aliases: |
CDA1, CDAI, CDAN1A, DLT, PRO1295 |
Location: |
Chr.15:42723557-42737219 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDAN1 | NM_138477.2 | 28 | 3981 | NP_612486.2 |
XM_005254176.4 | 28 | 3896 | XP_005254233.1 | |
XM_011521270.2 | 28 | 3921 | XP_011519572.1 | |
XM_011521271.2 | 28 | 3918 | XP_011519573.1 | |
XM_011521274.2 | 25 | 2967 | XP_011519576.1 | |
AF525398.1 | 28 | 3978 | AAO14994.1 | |
AL133019.1 | 1 | 1327 | ||
BC001092.2 | 20 | 2436 | AAH01092.1 | |
BC008333.1 | 20 | 2436 | AAH08333.1 | |
BC008334.1 | 20 | 2436 | AAH08334.1 | |
BC052568.1 | 28 | 3869 | AAH52568.1 | |
BC066640.1 | 28 | 3869 | AAH66640.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528725 | Chr.15:42463357 - 42812208 on Build GRCh38 | Gain | TTBK2 HAUS2 CDAN1 SNAP23 LRRC57 STARD9 ZNF106 |
nsv952615 | Chr.15:42714903 - 42736802 on Build GRCh38 | Deletion | CDAN1 STARD9 |
nsv457122 | Chr.15:42656664 - 42770350 on Build GRCh38 | Loss | TTBK2 CDAN1 STARD9 |
nsv517553 | Chr.15:42678701 - 42731284 on Build GRCh38 | Gain+Loss | CDAN1 STARD9 |
Set Membership: |
Intragenic Exonic DGV Variation |