Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55135 |
Gene Name: | WD repeat containing antisense to TP53 |
Gene Aliases: |
DKCB3, TCAB1, WDR79 |
Location: |
Chr.17:7686071-7703502 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WRAP53 | NM_001143990.1 | NP_001137462.1 | ||
NM_001143991.1 | NP_001137463.1 | |||
NM_001143992.1 | NP_001137464.1 | |||
NM_018081.2 | NP_060551.2 | |||
XM_011523952.2 | XP_011522254.1 | |||
XM_017024812.1 | XP_016880301.1 | |||
AK001247.1 | BAA91579.1 | |||
AK056669.1 | ||||
AK092370.1 | ||||
AK225273.1 | ||||
AK225411.1 | ||||
AK303246.1 | ||||
AY766322.1 | ||||
BC002336.2 | AAH02336.1 | |||
DQ431240.1 | ||||
DQ431241.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952119 | Chr.17:7700483 - 7709982 on Build GRCh38 | Deletion | EFNB3 WRAP53 |
Set Membership: |
Intragenic Non-exonic DGV Variation |