Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5889 |
Gene Name: | RAD51 paralog C |
Gene Aliases: |
BROVCA3, FANCO, R51H3, RAD51L2 |
Location: |
Chr.17:58692538-58734342 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAD51C | NM_002876.3 | 1 | 74 | NP_002867.1 |
NM_058216.2 | 1 | 74 | NP_478123.1 | |
NR_103872.1 | 1 | 74 | ||
NR_103873.1 | ||||
XM_006722001.3 | XP_006722064.1 | |||
XM_006722002.3 | XP_006722065.1 | |||
XM_006722004.3 | XP_006722067.1 | |||
XM_006722005.3 | XP_006722068.1 | |||
XM_017024914.1 | XP_016880403.1 | |||
XM_017024916.1 | XP_016880405.1 | |||
XM_017024917.1 | XP_016880406.1 | |||
AA304618.1 | ||||
AF029669.1 | AAC39604.1 | |||
AF029670.1 | AAC39605.1 | |||
AI424321.1 | ||||
AK129738.1 | ||||
AK290487.1 | ||||
AK303363.1 | ||||
BC093930.1 | AAH93930.1 | |||
BC101485.1 | AAI01486.1 | |||
BC107753.1 | 1 | 78 | AAI07754.1 | |
BT007339.1 | AAP36003.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833498 | Chr.17:58652063 - 58815031 on Build GRCh38 | Loss | TEX14 RAD51C PPM1E |
nsv1066945 | Chr.17:58650965 - 58704810 on Build GRCh38 | Loss | TEX14 RAD51C |
nsv428347 | Chr.17:58565015 - 58717588 on Build GRCh38 | Gain | TEX14 RAD51C |
Set Membership: |
Intragenic Exonic DGV Variation |