Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8518 |
Gene Name: | inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein |
Gene Aliases: |
DYS, ELP1, FD, IKAP, IKI3, TOT1 |
Location: |
Chr.9:108867520-108934332 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IKBKAP | NM_001318360.1 | NP_001305289.1 | ||
NM_003640.4 | NP_003631.2 | |||
XM_011519136.1 | XP_011517438.1 | |||
XM_011519137.1 | XP_011517439.1 | |||
XM_017015238.1 | XP_016870727.1 | |||
AB210033.1 | ||||
AF044195.1 | AAC64258.1 | |||
AF153419.2 | AAG43369.1 | |||
AK001641.1 | ||||
AK022559.1 | ||||
AK024223.1 | ||||
AK123257.1 | ||||
AK127237.1 | ||||
AK225798.1 | ||||
AK289962.1 | ||||
AK304742.1 | ||||
AL049945.1 | CAB43219.1 | |||
BC033094.1 | AAH33094.1 | |||
CR749385.1 | CAH18235.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6659 | Chr.9:108856263 - 108890282 on Build GRCh38 | Insertion | ACTL7A IKBKAP |
Set Membership: |
Intragenic Non-exonic DGV Variation |