Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 51082 |
Gene Name: | RNA polymerase I subunit D |
Gene Aliases: |
AC19, POLR1C, RPA16, RPA9, RPAC2, RPC16, RPO1-3, TCS2 |
Location: |
Chr.13:27620743-27667422 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLR1D | NM_001206559.1 | NP_001193488.1 | ||
NM_015972.3 | 2 | 691 | NP_057056.1 | |
NM_152705.2 | NP_689918.1 | |||
XM_005266412.1 | 3 | 1531 | XP_005266469.1 | |
XM_005266414.2 | XP_005266471.1 | |||
XM_017020623.1 | XP_016876112.1 | |||
AF077044.1 | 2 | 579 | AAD27777.1 | |
AK000623.1 | 2 | 577 | ||
AK097973.1 | ||||
AK307356.1 | ||||
AK311833.1 | ||||
BC000889.1 | 2 | 595 | AAH00889.1 | |
BC006972.1 | AAH06972.1 | |||
BC015319.2 | AAH15319.1 | |||
BC018528.1 | AAH18528.1 | |||
BI253733.1 | 2 | 503 | ||
DB123903.1 | ||||
DB455307.1 | ||||
DB459107.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1039183 | Chr.13:27600204 - 27632386 on Build GRCh38 | Loss | LNX2 POLR1D |
Set Membership: |
Intragenic Exonic DGV Variation |