Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 10166 |
Gene Name: | solute carrier family 25 member 15 |
Gene Aliases: |
D13S327, HHH, ORC1, ORNT1 |
Location: |
Chr.13:40789411-40812460 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A15 | NM_014252.3 | NP_055067.1 | ||
AF112968.1 | AAD45238.1 | |||
AF177333.1 | AAG17977.1 | |||
AK296862.1 | ||||
AK313602.1 | ||||
AL538057.3 | ||||
BC002702.2 | AAH02702.1 | |||
BI461478.1 |
Entrez Gene ID: | 100616668 |
Gene Name: | transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 5 |
Gene Aliases: |
- |
Location: |
Chr.13:40796985-40921750 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPTE2P5 | NR_038258.1 | 8 | 2047 | |
NR_038259.1 | 6 | 1876 | ||
AK093232.1 | ||||
AK096993.1 | 6 | 1864 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892332 | Chr.13:40619338 - 40820761 on Build GRCh38 | Gain | MIR320D1 FOXO1 MRPS31 SLC25A15 TPTE2P5 MIR621 |
Set Membership: |
Intragenic Exonic Non-exonic DGV Variation |