Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 11315 |
Gene Name: | Parkinsonism associated deglycase |
Gene Aliases: |
DJ-1, DJ1, HEL-S-67p |
Location: |
Chr.1:7961654-7985282 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PARK7 | NM_001123377.1 | 7 | 787 | NP_001116849.1 |
NM_007262.4 | 7 | 845 | NP_009193.2 | |
XM_005263424.3 | 7 | 865 | XP_005263481.1 | |
AF021819.1 | 7 | 753 | AAC12806.1 | |
AK091679.1 | 1 | 2208 | ||
AK130886.1 | 6 | 680 | ||
BC008188.2 | 7 | 766 | AAH08188.1 | |
D61380.2 | 6 | 688 | BAA09603.2 | |
EU794641.1 | 6 | 688 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1521 | Chr.1:7954179 - 7987433 on Build GRCh38 | Insertion | PARK7 |
Set Membership: |
Intragenic Exonic DGV Variation |