Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 84464 |
Gene Name: | SLX4 structure-specific endonuclease subunit |
Gene Aliases: |
BTBD12, FANCP, MUS312 |
Location: |
Chr.16:3581181-3611598 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLX4 | NM_032444.2 | 12 | 4809 | NP_115820.2 |
XM_011522715.2 | 12 | 4823 | XP_011521017.1 | |
XM_017023775.1 | 9 | 4214 | XP_016879264.1 | |
AK095411.1 | 1 | 679 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833128 | Chr.16:3525122 - 3690497 on Build GRCh38 | Loss | NLRC3 LOC101929732 DNASE1 SLX4 CLUAP1 TRAP1 |
nsv518509 | Chr.16:3583656 - 3688960 on Build GRCh38 | Gain | DNASE1 SLX4 TRAP1 |
nsv517451 | Chr.16:3549655 - 3611814 on Build GRCh38 | Loss | NLRC3 LOC101929732 DNASE1 SLX4 |
Set Membership: |
Intragenic Exonic DGV Variation |