Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 301 |
Gene Name: | annexin A1 |
Gene Aliases: |
ANX1, LPC1 |
Location: |
Chr.9:73151731-73170393 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANXA1 | NM_000700.2 | NP_000691.1 | ||
XM_011518609.1 | XP_011516911.1 | |||
XM_017014657.1 | XP_016870146.1 | |||
AA039737.1 | ||||
AK074480.1 | ||||
BC001275.1 | AAH01275.1 | |||
BC034157.1 | ||||
BC035993.1 | AAH35993.1 | |||
X05908.1 | CAA29338.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3620660 | Chr.9:73148689 - 73332974 on Build GRCh38 | Loss | ANXA1 |
nsv6561 | Chr.9:73140727 - 73175683 on Build GRCh38 | Insertion | ANXA1 |
esv3620662 | Chr.9:73155774 - 73254536 on Build GRCh38 | Gain | ANXA1 |
nsv526225 | Chr.9:72987987 - 73372263 on Build GRCh38 | Gain | ANXA1 LOC105376081 |
nsv614612 | Chr.9:72979640 - 73296530 on Build GRCh38 | Gain | ANXA1 LOC105376081 |
Set Membership: |
Intragenic Non-exonic DGV Variation |