Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 25978 |
Gene Name: | charged multivesicular body protein 2B |
Gene Aliases: |
ALS17, CHMP2.5, DMT1, VPS2-2, VPS2B |
Location: |
Chr.3:87227263-87255548 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHMP2B | NM_001244644.1 | 5 | 2129 | NP_001231573.1 |
NM_014043.3 | 6 | 2221 | NP_054762.2 | |
XM_011533576.2 | 6 | 2052 | XP_011531878.1 | |
AK002165.1 | 6 | 2194 | ||
BC001553.1 | 6 | 2037 | AAH01553.1 | |
BM982148.1 | 1 | 423 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3568885 | Chr.3:87182571 - 87518398 on Build GRCh38 | Loss | CHMP2B MIR4795 POU1F1 |
nsv527279 | Chr.3:86796784 - 87814130 on Build GRCh38 | Gain | LINC00506 HTR1F CHMP2B VGLL3 MIR4795 POU1F1 |
Set Membership: |
Intragenic Exonic DGV Variation |