Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 255239 |
Gene Name: | ankyrin repeat and kinase domain containing 1 |
Gene Aliases: |
PKK2 |
Location: |
Chr.11:113386014-113400418 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANKK1 | NM_178510.1 | NP_848605.1 | ||
XM_011542736.2 | XP_011541038.1 | |||
XM_011542737.2 | XP_011541039.1 | |||
XM_011542738.2 | XP_011541040.1 | |||
XM_017017475.1 | XP_016872964.1 | |||
AF487542.1 | AAQ09005.1 | |||
AJ541797.2 | CAD62569.2 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483080 | Chr.11:113216072 - 113400474 on Build GRCh38 | Loss | ANKK1 TTC12 NCAM1 NCAM1-AS1 |
nsv1040152 | Chr.11:113387343 - 113415194 on Build GRCh38 | Loss | DRD2 ANKK1 |
nsv556444 | Chr.11:113318201 - 113512672 on Build GRCh38 | Gain | DRD2 ANKK1 MIR4301 TTC12 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |