Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 284058 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 |
Gene Aliases: |
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1 |
Location: |
Chr.17:46029916-46225374 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KANSL1 | NM_001193465.1 | 16 | 3821 | NP_001180394.1 |
NM_001193466.1 | 15 | 4038 | NP_001180395.1 | |
NM_015443.3 | 15 | 3740 | NP_056258.1 | |
XM_006721823.1 | 16 | 3824 | XP_006721886.1 | |
XM_006721824.3 | 16 | 4008 | XP_006721887.1 | |
XM_011524628.2 | 16 | 4005 | XP_011522930.1 | |
XM_011524629.2 | 15 | 3906 | XP_011522931.1 | |
XM_011524630.2 | 15 | 3819 | XP_011522932.1 | |
XM_011524631.2 | 15 | 3817 | XP_011522933.1 | |
XM_011524632.2 | 15 | 2434 | XP_011522934.1 | |
AB033093.1 | 14 | 3661 | BAA86581.1 | |
AK094946.1 | 7 | 1539 | ||
AK291259.1 | 15 | 3798 | ||
AK310569.1 | 1 | 1340 | ||
AL117476.1 | 4 | 908 | CAB55949.1 | |
AL137317.1 | 11 | 1982 | CAB70694.1 | |
BC035892.1 | 10 | 1845 | AAH35892.1 | |
BC098376.1 | 15 | 3740 | AAH98376.1 | |
BX538006.1 | 15 | 4041 | CAD97958.1 | |
BX648760.1 | 8 | 7798 | CAH10565.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471698 | Chr.17:46002113 - 46300887 on Build GRCh38 | Gain+Loss | MAPT ARL17B KANSL1-AS1 KANSL1 LRRC37A |
nsv469705 | Chr.17:45969304 - 46127121 on Build GRCh38 | Loss | MAPT STH KANSL1 |
nsv469537 | Chr.17:46002112 - 46163353 on Build GRCh38 | Gain+Loss | MAPT KANSL1 |
nsv575142 | Chr.17:46003698 - 46172742 on Build GRCh38 | Loss | MAPT KANSL1 |
esv2762433 | Chr.17:45496768 - 46710828 on Build GRCh38 | Gain+Loss | MGC57346 NSF LRRC37A2 SPPL2C MAPT LOC107987254 MGC57346-CRHR1 CRHR1-IT1 LOC107987253 LOC644172 LRRC37A4P NSFP1 KANSL1-AS1 STH KANSL1 LRRC37A ARL17B LOC105376839 MAPT-AS1 CRHR1 MAPT-IT1 ARL17A |
nsv1072740 | Chr.17:45970433 - 46248034 on Build GRCh38 | Deletion | MAPT KANSL1-AS1 STH KANSL1 |
dgv3178n100 | Chr.17:45877004 - 46034461 on Build GRCh38 | Gain | MAPT STH MAPT-AS1 MAPT-IT1 KANSL1 |
esv2751686 | Chr.17:45962198 - 46286690 on Build GRCh38 | Gain | MAPT ARL17B KANSL1-AS1 STH KANSL1 LRRC37A |
esv2758692 | Chr.17:46006548 - 47199967 on Build GRCh38 | Gain+Loss | WNT9B NSF CDC27 LRRC37A2 MAPT LOC107987254 LOC107987253 NSFP1 KANSL1-AS1 RPRML KANSL1 LRRC37A WNT3 RPS2P47 MIR5089 ARL17B GOSR2 ARL17A |
dgv128e55 | Chr.17:46017105 - 46286690 on Build GRCh38 | Gain | MAPT ARL17B KANSL1-AS1 KANSL1 LRRC37A |
dgv127e55 | Chr.17:45927606 - 46733750 on Build GRCh38 | Gain | NSF LRRC37A2 MAPT LOC107987254 ARL17B LOC107987253 NSFP1 KANSL1-AS1 STH KANSL1 ARL17A LRRC37A |
Set Membership: |
Intragenic Exonic DGV Variation |