Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 7133 |
Gene Name: | TNF receptor superfamily member 1B |
Gene Aliases: |
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR |
Location: |
Chr.1:12166948-12209222 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNFRSF1B | NM_001066.2 | NP_001057.1 | ||
XM_011542060.2 | XP_011540362.1 | |||
XM_011542063.2 | XP_011540365.1 | |||
XM_017002211.1 | XP_016857700.1 | |||
XM_017002212.1 | XP_016857701.1 | |||
XM_017002213.1 | XP_016857702.1 | |||
XM_017002214.1 | XP_016857703.1 | |||
XM_017002215.1 | XP_016857704.1 | |||
AY148473.1 | AAN72434.1 | |||
BC011844.2 | AAH11844.1 | |||
BC042167.1 | AAH42167.1 | |||
BC052977.1 | AAH52977.1 | |||
BT019927.1 | AAV38730.1 | |||
EU826615.1 | ||||
M32315.1 | AAA59929.1 | |||
M55994.1 | AAA36755.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471622 | Chr.1:12155207 - 12371588 on Build GRCh38 | Gain+Loss | TNFRSF1B MIR7846 VPS13D MIR4632 |
nsv508891 | Chr.1:12097920 - 12235045 on Build GRCh38 | Insertion | TNFRSF1B MIR7846 VPS13D MIR4632 TNFRSF8 |
esv3584475 | Chr.1:12134394 - 12217607 on Build GRCh38 | Gain | TNFRSF1B MIR7846 MIR4632 TNFRSF8 |
nsv527024 | Chr.1:12164182 - 12207208 on Build GRCh38 | Loss | TNFRSF1B MIR7846 MIR4632 |
nsv469830 | Chr.1:12039669 - 12199858 on Build GRCh38 | Loss | TNFRSF1B MIR7846 MIR4632 TNFRSF8 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |