Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55063 |
Gene Name: | zinc finger CW-type and PWWP domain containing 1 |
Gene Aliases: |
ZCW1 |
Location: |
Chr.7:100400826-100429239 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZCWPW1 | NM_001258008.1 | NP_001244937.1 | ||
NM_017984.4 | NP_060454.3 | |||
XM_005250479.2 | XP_005250536.1 | |||
XM_005250480.2 | XP_005250537.1 | |||
XM_006716035.3 | XP_006716098.1 | |||
XM_006716036.2 | XP_006716099.1 | |||
XM_006716037.3 | XP_006716100.1 | |||
XM_006716038.3 | XP_006716101.1 | |||
XM_006716040.3 | XP_006716103.1 | |||
XM_017012379.1 | XP_016867868.1 | |||
XM_017012380.1 | XP_016867869.1 | |||
AK093038.1 | BAC04028.1 | |||
AK300745.1 | ||||
AK302108.1 | ||||
AL136735.1 | CAB66669.1 | |||
BC002725.1 | AAH02725.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | MIR93 EPO VGF MIR6840 EPHB4 MUC12 SERPINE1 STAG3L5P AP1S1 GAL3ST4 SRRT AZGP1 MBLAC1 MUC17 ZAN NAT16 STAG3 TFR2 LOC102724094 C7orf43 MIR4658 TAF6 SPDYE3 MIR6875 LRCH4 GATS GPC2 GNB2 ZKSCAN1 ACTL6B PVRIG2P UFSP1 CNPY4 FBXO24 AZGP1P1 ZSCAN21 SAP25 PILRA AGFG2 ZNHIT1 FIS1 PVRIG MEPCE MUC3A CLDN15 MOSPD3 MCM7 POP7 ZNF3 TSC22D4 MIR4653 PMS2P1 C7orf61 PLOD3 ACHE MOGAT3 LOC105375429 NYAP1 MIR25 PILRB MIR106B TRIP6 TRIM56 STAG3L5P-PVRIG2P-PILRB COPS6 AP4M1 PCOLCE ZCWPW1 PPP1R35 LAMTOR4 SLC12A9 ZASP PCOLCE-AS1 GIGYF1 |
nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss | MEPCE NYAP1 PILRB MIR6840 STAG3L5P-PVRIG2P-PILRB SPDYE3 STAG3L5P TSC22D4 ZCWPW1 PMS2P1 PVRIG2P C7orf61 PPP1R35 PILRA |
nsv527107 | Chr.7:100417088 - 100467096 on Build GRCh38 | Loss | MEPCE C7orf61 PPP1R35 TSC22D4 ZCWPW1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |