Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5889 |
Gene Name: | RAD51 paralog C |
Gene Aliases: |
BROVCA3, FANCO, R51H3, RAD51L2 |
Location: |
Chr.17:58692538-58734342 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833498 | Chr.17:58652063 - 58815031 on Build GRCh38 | Loss | TEX14 RAD51C PPM1E |
nsv1066945 | Chr.17:58650965 - 58704810 on Build GRCh38 | Loss | TEX14 RAD51C |
nsv428347 | Chr.17:58565015 - 58717588 on Build GRCh38 | Gain | TEX14 RAD51C |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |