Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 90332 |
Gene Name: | exocyst complex component 3 like 2 |
Gene Aliases: |
XTP7 |
Location: |
Chr.19:45212621-45234211 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L2 | NM_138568.3 | NP_612635.3 | ||
AK093466.1 | BAC04175.1 | |||
AK225791.1 | ||||
BC104947.1 | AAI04948.1 | |||
BC104949.1 | AAI04950.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422482 | Chr.19:45173447 - 45266547 on Build GRCh38 | Duplication | EXOC3L2 MARK4 TRAPPC6A BLOC1S3 |
esv3583434 | Chr.19:45185375 - 45248399 on Build GRCh38 | Loss | EXOC3L2 BLOC1S3 |
nsv833844 | Chr.19:45181051 - 45384148 on Build GRCh38 | Loss | PPP1R13L CKM EXOC3L2 MARK4 KLC3 ERCC2 BLOC1S3 |
nsv833843 | Chr.19:45090630 - 45290424 on Build GRCh38 | Loss | EXOC3L2 GEMIN7 MARK4 PPP1R37 NKPD1 TRAPPC6A BLOC1S3 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |