Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 50943 |
Gene Name: | forkhead box P3 |
Gene Aliases: |
AIID, DIETER, IPEX, JM2, PIDX, XPID |
Location: |
Chr.X:49250436-49266505 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | NP_001107849.1 | ||
NM_014009.3 | NP_054728.2 | |||
XM_006724533.2 | XP_006724596.2 | |||
XM_011543916.2 | XP_011542218.1 | |||
XM_017029565.1 | XP_016885054.1 | |||
XM_017029566.1 | XP_016885055.1 | |||
XM_017029567.1 | XP_016885056.1 | |||
AF277993.1 | AAG53607.1 | |||
AJ005891.1 | CAA06748.1 | |||
AK292052.1 | ||||
AK299988.1 | ||||
BC113401.1 | ||||
BC113403.1 | ||||
BC143785.1 | ||||
BC143786.1 | ||||
DQ010327.1 | AAY27088.1 | |||
EF534714.1 | ||||
EU855812.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain | CACNA1F GAGE10 CCDC22 FOXP3 PPP1R3F |
nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain | PORCN OTUD5 CCDC120 SUV39H1 GATA1 CACNA1F SYP-AS1 EBP KCND1 CCDC22 TFE3 PQBP1 GPKOW RBM3 GRIPAP1 WDR45 SYP PRICKLE3 PCSK1N PLP2 FOXP3 MAGIX WDR13 TBC1D25 PRAF2 LOC101927635 FTSJ1 WAS PIM2 SLC35A2 PPP1R3F HDAC6 ERAS TIMM17B GLOD5 SLC38A5 |
esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss | PORCN OTUD5 CCDC120 SSX3 SUV39H1 GATA1 CACNA1F SYP-AS1 EBP KCND1 CCDC22 TFE3 PQBP1 GPKOW RBM3 GRIPAP1 WDR45 SYP PRICKLE3 PCSK1N PLP2 ZNF182 FOXP3 MAGIX SSX4 ZNF630-AS1 WDR13 TBC1D25 PRAF2 SSX1 LOC101927635 SSX6 FTSJ1 WAS SPACA5B PIM2 SLC35A2 PPP1R3F HDAC6 ERAS ZNF630 SSX9 SSX4B TIMM17B SPACA5 GLOD5 SLC38A5 SSX5 |
nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss | CACNA1F GAGE10 CCDC22 FOXP3 PPP1R3F |
nsv6895 | Chr.X:49227319 - 49262690 on Build GRCh38 | Insertion | CACNA1F CCDC22 FOXP3 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |