Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 60506 |
Gene Name: | nyctalopin |
Gene Aliases: |
CLRP, CSNB1, CSNB1A, CSNB4, NBM1 |
Location: |
Chr.X:41447460-41476414 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NYX | NM_022567.2 | NP_072089.1 | ||
XM_017029709.1 | XP_016885198.1 | |||
AF254868.1 | AAG42685.1 | |||
AJ278865.1 | CAC19014.1 | |||
BC112242.1 | AAI12243.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508763 | Chr.X:41415959 - 41531609 on Build GRCh38 | Insertion | NYX CASK |
esv33720 | Chr.X:40891487 - 43157518 on Build GRCh38 | Loss | GPR82 USP9X DDX3X GPR34 NYX LOC107985678 LOC105373185 LOC643043 PPP1R2P9 CASK |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |