Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2780 |
Gene Name: | G protein subunit alpha transducin 2 |
Gene Aliases: |
ACHM4, GNATC |
Location: |
Chr.1:109603091-109619733 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNAT2 | XM_011541264.2 | XP_011539566.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2299 | Chr.1:109610296 - 109662341 on Build GRCh38 | Deletion | GSTM4 AMPD2 GNAT2 |
dgv253n100 | Chr.1:109613590 - 109700319 on Build GRCh38 | Gain | GSTM2 GSTM1 GSTM4 AMPD2 GNAT2 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |