Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 79188 |
Gene Name: | transmembrane protein 43 |
Gene Aliases: |
ARVC5, ARVD5, EDMD7, LUMA |
Location: |
Chr.3:14124940-14143680 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM43 | NM_024334.2 | NP_077310.1 | ||
XM_017007176.1 | XP_016862665.1 | |||
AK027466.1 | BAB55131.1 | |||
AK027757.1 | BAB55348.1 | |||
AK027827.1 | BAB55396.1 | |||
AK027877.1 | BAB55425.1 | |||
AK074073.1 | 9 | 3191 | BAB84899.1 | |
AK075010.1 | BAC11350.1 | |||
AK225589.1 | ||||
AL136916.1 | CAB66850.1 | |||
AY358625.1 | AAQ88988.1 | |||
BC003125.1 | AAH03125.1 | |||
BC008054.2 | AAH08054.2 | |||
BC011719.2 | AAH11719.1 | |||
KU178769.1 | ||||
KU178770.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1011766 | Chr.3:13485961 - 14213016 on Build GRCh38 | Gain | FGD5P1 TPRXL SNORA93 LOC100293612 LINC00620 WNT7A LSM3 XPC HDAC11 TMEM43 FBLN2 CHCHD4 |
nsv521096 | Chr.3:14135751 - 14154182 on Build GRCh38 | Loss | XPC TMEM43 |
esv34163 | Chr.3:13865660 - 14212447 on Build GRCh38 | Loss | FGD5P1 TPRXL WNT7A LSM3 XPC TMEM43 CHCHD4 |
esv2724945 | Chr.3:13931758 - 14820330 on Build GRCh38 | Deletion | TPRXL SLC6A6 LOC152274 GRIP2 TMEM43 FGD5P1 LINC01267 CCDC174 LSM3 XPC FGD5 C3orf20 CHCHD4 |
nsv955318 | Chr.3:14126901 - 14142100 on Build GRCh38 | Deletion | TMEM43 |
nsv589769 | Chr.3:14136822 - 14139133 on Build GRCh38 | Loss | TMEM43 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |