Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4684 |
Gene Name: | neural cell adhesion molecule 1 |
Gene Aliases: |
CD56, MSK39, NCAM |
Location: |
Chr.11:112961247-113278436 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCAM1 | NM_000615.6 | NP_000606.3 | ||
NM_001076682.3 | NP_001070150.1 | |||
NM_001242607.1 | NP_001229536.1 | |||
NM_001242608.1 | NP_001229537.1 | |||
NM_181351.4 | NP_851996.2 | |||
AB209443.1 | BAD92680.1 | |||
AF005070.1 | AAB61615.1 | |||
AK292453.1 | ||||
AK314589.1 | ||||
BC029119.1 | ||||
BC047244.1 | AAH47244.1 | |||
DA231540.1 | ||||
DA337649.1 | ||||
S71824.1 | AAB31836.1 | |||
U63041.1 | AAB04558.1 | |||
X16841.1 | CAA34739.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951071 | Chr.11:112964879 - 112968078 on Build GRCh38 | Duplication | NCAM1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |