Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 54982 |
Gene Name: | ceroid-lipofuscinosis, neuronal 6, late infantile, variant |
Gene Aliases: |
CLN4A, HsT18960, nclf |
Location: |
Chr.15:68206992-68229742 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLN6 | NM_017882.2 | NP_060352.1 | ||
AK000568.1 | BAA91260.1 | |||
AK027604.1 | BAB55226.1 | |||
AK074753.1 | BAC11181.1 | |||
AK291175.1 | ||||
AK293155.1 | ||||
AK293197.1 | ||||
BC010849.1 | AAH10849.1 | |||
BC013130.1 | AAH13130.1 | |||
CR457244.1 | CAG33525.1 | |||
KU178671.1 | ||||
KU178672.1 | ||||
KU178673.1 | ||||
KU178674.1 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |