Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5604 |
Gene Name: | mitogen-activated protein kinase kinase 1 |
Gene Aliases: |
CFC3, MAPKK1, MEK1, MKK1, PRKMK1 |
Location: |
Chr.15:66386873-66491544 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAP2K1 | NM_002755.3 | NP_002746.1 | ||
XM_011521783.2 | XP_011520085.1 | |||
XM_017022411.1 | XP_016877900.1 | |||
XM_017022412.1 | XP_016877901.1 | |||
AK291500.1 | ||||
AK294322.1 | ||||
AK312356.1 | ||||
BC137459.1 | ||||
BC139729.1 | ||||
L05624.1 | AAA36318.1 | |||
L11284.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833045 | Chr.15:66295248 - 66460419 on Build GRCh38 | Gain | MAP2K1 TIPIN DIS3L SCARNA14 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |