Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3990 |
Gene Name: | lipase C, hepatic type |
Gene Aliases: |
HDLCQ12, HL, HTGL, LIPH |
Location: |
Chr.15:58410754-58568952 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPC | XM_005254372.1 | XP_005254429.1 | ||
AK290062.1 | ||||
AK292631.1 | ||||
BC146659.1 |
Entrez Gene ID: | 101928635 |
Gene Name: | uncharacterized LOC101928635 |
Gene Aliases: |
- |
Location: |
Chr.15:58106609-58431770 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928635 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892705 | Chr.15:58414647 - 58428232 on Build GRCh38 | Loss | LIPC LOC101928635 |
nsv569598 | Chr.15:58260407 - 58560264 on Build GRCh38 | Loss | LIPC LOC101928635 LIPC-AS1 LOC101928668 |
nsv1047048 | Chr.15:58409201 - 58492601 on Build GRCh38 | Loss | LIPC LOC101928635 LIPC-AS1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |