Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 161742 |
Gene Name: | sprouty related EVH1 domain containing 1 |
Gene Aliases: |
NFLS, PPP1R147, hSpred1, spred-1 |
Location: |
Chr.15:38252087-38357249 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRED1 | NM_152594.2 | NP_689807.1 | ||
XM_005254202.3 | XP_005254259.1 | |||
XM_011521289.2 | XP_011519591.1 | |||
AK091222.1 | BAC03614.1 | |||
AY299089.1 | AAP59414.1 | |||
BC018015.1 | ||||
BC137480.1 | ||||
BC137481.1 | ||||
DA405650.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892674 | Chr.15:37311235 - 38400599 on Build GRCh38 | Gain | LOC101928227 SPRED1 TMCO5A |
nsv832974 | Chr.15:38206405 - 38398384 on Build GRCh38 | Loss | SPRED1 |
nsv832973 | Chr.15:38151604 - 38352681 on Build GRCh38 | Gain | SPRED1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |