Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55737 |
Gene Name: | VPS35 retromer complex component |
Gene Aliases: |
MEM3, PARK17 |
Location: |
Chr.16:46657979-46689232 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VPS35 | NM_018206.5 | NP_060676.2 | ||
XM_005256045.3 | XP_005256102.1 | |||
XM_011523227.2 | XP_011521529.1 | |||
AF175265.1 | AAF89953.1 | |||
AF186382.1 | AAG40619.1 | |||
AF191298.2 | AAF02778.2 | |||
AK001614.1 | BAA91790.1 | |||
AK223218.1 | BAD96938.1 | |||
AL136888.1 | CAB66822.1 | |||
BC010362.1 | AAH10362.1 | |||
BC093036.1 | AAH93036.1 | |||
DB049686.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv572536 | Chr.16:46660737 - 46693473 on Build GRCh38 | Loss | VPS35 ORC6 |
nsv526656 | Chr.16:46505480 - 46812751 on Build GRCh38 | Gain | VPS35 MYLK3 ORC6 C16orf87 ANKRD26P1 SHCBP1 |
dgv2975n100 | Chr.16:46429858 - 46768065 on Build GRCh38 | Gain | VPS35 MYLK3 ORC6 ANKRD26P1 SHCBP1 |
esv3571142 | Chr.16:46430577 - 46840772 on Build GRCh38 | Gain | VPS35 MYLK3 ORC6 C16orf87 ANKRD26P1 SHCBP1 |
esv3638540 | Chr.16:46678001 - 46738780 on Build GRCh38 | Gain | VPS35 MYLK3 ORC6 |
esv3892855 | Chr.16:46671431 - 46750308 on Build GRCh38 | Loss | VPS35 MYLK3 ORC6 |
esv3638534 | Chr.16:46547097 - 46716009 on Build GRCh38 | Gain | VPS35 MYLK3 ORC6 ANKRD26P1 SHCBP1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |