Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8301 |
Gene Name: | phosphatidylinositol binding clathrin assembly protein |
Gene Aliases: |
CALM, CLTH, LAP |
Location: |
Chr.11:85957171-86069881 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PICALM | NM_001008660.2 | 20 | 3938 | NP_001008660.1 |
NM_001206946.1 | 20 | 4043 | NP_001193875.1 | |
NM_001206947.1 | 19 | 3629 | NP_001193876.1 | |
NM_007166.3 | 20 | 4064 | NP_009097.2 | |
AK128786.1 | 1 | 1792 | ||
CA415536.1 | 1 | 106 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1053808 | Chr.11:85877876 - 86046651 on Build GRCh38 | Gain | CCDC83 PICALM |
nsv1035489 | Chr.11:85791910 - 86067187 on Build GRCh38 | Gain | SYTL2 CCDC83 PICALM |
dgv1247n100 | Chr.11:85930576 - 85973892 on Build GRCh38 | Gain | PICALM |
Set Membership: |
Intragenic Exonic DGV Variation |