Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3312 |
Gene Name: | heat shock protein family A (Hsp70) member 8 |
Gene Aliases: |
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71 |
Location: |
Chr.11:123057492-123062366 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSPA8 | NM_006597.5 | NP_006588.1 | ||
NM_153201.3 | NP_694881.1 | |||
XM_011542798.1 | XP_011541100.1 | |||
AB034951.1 | BAB18615.1 | |||
AF352832.1 | AAK17898.1 | |||
AK096100.1 | ||||
AK129885.1 | ||||
AK222628.1 | BAD96348.1 | |||
AK222785.1 | BAD96505.1 | |||
AK300404.1 | ||||
AK303935.1 | ||||
AK310467.1 | ||||
BC016179.1 | AAH16179.1 | |||
BC016660.1 | AAH16660.1 | |||
BC019816.2 | AAH19816.1 | |||
DC312904.1 | ||||
DC395565.1 | ||||
EU668349.1 | ||||
FJ224294.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1039284 | Chr.11:122985952 - 123083303 on Build GRCh38 | Gain | CLMP SNORD14C HSPA8 SNORD14D LOC341056 SNORD14E |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |