Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 79621 |
Gene Name: | ribonuclease H2 subunit B |
Gene Aliases: |
AGS2, DLEU8 |
Location: |
Chr.13:50909678-50970462 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RNASEH2B | NM_001142279.2 | NP_001135751.1 | ||
NM_024570.3 | NP_078846.2 | |||
XM_006719867.3 | XP_006719930.1 | |||
XM_011535229.1 | XP_011533531.1 | |||
XM_011535230.2 | XP_011533532.1 | |||
XM_011535231.2 | XP_011533533.1 | |||
XM_011535233.2 | XP_011533535.1 | |||
XM_017020747.1 | XP_016876236.1 | |||
AK021774.1 | BAB13892.1 | |||
AK223340.1 | BAD97060.1 | |||
AY764036.1 | AAX13343.1 | |||
BC001397.2 | AAH01397.1 | |||
BC005088.1 | ||||
BC007332.2 | AAH07332.1 | |||
BC010174.2 | AAH10174.1 | |||
BC036744.1 | AAH36744.1 | |||
BX382357.2 | ||||
DB221997.1 | ||||
DT216141.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss | DLEU2 MIR3613 KCNRG GUCY1B2 DLEU7-AS1 FAM124A INTS6 DLEU1-AS1 INTS6-AS1 LINC00371 ST13P4 RNASEH2B MIR16-1 MIR15A DLEU7 TRIM13 MIR5693 SPRYD7 DLEU1 RNASEH2B-AS1 SERPINE3 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |