Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 203228 |
Gene Name: | chromosome 9 open reading frame 72 |
Gene Aliases: |
ALSFTD, FTDALS, FTDALS1 |
Location: |
Chr.9:27546546-27573866 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C9orf72 | NM_001256054.2 | NP_001242983.1 | ||
NM_018325.4 | NP_060795.1 | |||
AK291425.1 | ||||
AL832229.1 | ||||
AL832467.1 | 6 | 1842 | ||
BC068445.1 | AAH68445.1 | |||
JN681271.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1015259 | Chr.9:26347305 - 27643043 on Build GRCh38 | Gain | IFT74 MOB3B LRRC19 TEK C9orf72 LINC00032 IFNK PLAA EQTN IFT74-AS1 CAAP1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |