Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55812 |
Gene Name: | spermatogenesis associated 7 |
Gene Aliases: |
HEL-S-296, HSD-3.1, HSD3, LCA3 |
Location: |
Chr.14:88385638-88470350 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv565512 | Chr.14:88427866 - 88573973 on Build GRCh38 | Gain | SPATA7 ZC3H14 PTPN21 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |