Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 1120 |
Gene Name: | choline kinase beta |
Gene Aliases: |
CHETK, CHKL, CK, CKB, CKEKB, EK, EKB, MDCMC |
Location: |
Chr.22:50578958-50582999 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHKB | NM_005198.4 | NP_005189.2 | ||
AB029886.1 | BAA82512.1 | |||
AK097775.1 | 3 | 1075 | ||
AK314324.1 | ||||
AL096780.1 | CAB46629.1 | |||
AL096781.1 | CAB46630.1 | |||
BC082263.1 | AAH82263.1 | |||
BC101488.1 | ||||
BC113521.1 | ||||
CR456419.1 | CAG30305.1 |
Entrez Gene ID: | 386593 |
Gene Name: | CHKB-CPT1B readthrough (NMD candidate) |
Gene Aliases: |
CHKL-CPT1B |
Location: |
Chr.22:50568861-50582999 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHKB-CPT1B | NR_027928.2 | |||
AB051457.1 | BAB33340.1 | |||
AK301477.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059234 | Chr.22:50560544 - 50617472 on Build GRCh38 | Gain | CHKB-AS1 SYCE3 CHKB MAPK8IP2 CPT1B CHKB-CPT1B |
nsv1057656 | Chr.22:50478783 - 50595520 on Build GRCh38 | Gain | LMF2 ODF3B NCAPH2 CPT1B TYMP CHKB-AS1 SYCE3 CHKB KLHDC7B ADM2 MIOX CHKB-CPT1B SCO2 |
nsv829340 | Chr.22:50505720 - 50603696 on Build GRCh38 | Gain | CHKB-AS1 SYCE3 CHKB KLHDC7B LMF2 ODF3B NCAPH2 MAPK8IP2 CPT1B TYMP CHKB-CPT1B SCO2 |
nsv834239 | Chr.22:50485317 - 50630215 on Build GRCh38 | Loss | LMF2 ODF3B NCAPH2 MAPK8IP2 CPT1B TYMP ARSA CHKB-AS1 SYCE3 CHKB KLHDC7B ADM2 MIOX CHKB-CPT1B SCO2 |
nsv955202 | Chr.22:50544472 - 50582171 on Build GRCh38 | Deletion | SYCE3 CHKB KLHDC7B CPT1B CHKB-CPT1B |
nsv589223 | Chr.22:50539595 - 50619495 on Build GRCh38 | Loss | CHKB-AS1 SYCE3 CHKB KLHDC7B MAPK8IP2 CPT1B CHKB-CPT1B |
nsv589222 | Chr.22:50491152 - 50589773 on Build GRCh38 | Gain | CHKB-AS1 SYCE3 CHKB KLHDC7B LMF2 ODF3B NCAPH2 CPT1B TYMP CHKB-CPT1B SCO2 |
Set Membership: |
Intragenic Non-exonic DGV Variation |