Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 18707 |
Gene Name: | phosphatidylinositol 3-kinase catalytic delta polypeptide |
Gene Aliases: |
2410099E07Rik, 2610208K16Rik, AW545373, p110delta |
Location: |
Chr.4:149649168-149702566 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pik3cd | NM_001029837.2 | 2 | 196 | NP_001025008.2 |
NM_001164049.1 | 2 | 223 | NP_001157521.1 | |
NM_001164050.1 | 2 | 223 | NP_001157522.1 | |
NM_001164051.1 | 2 | 247 | NP_001157523.1 | |
NM_008840.3 | 3 | 398 | NP_032866.2 | |
AK011923.1 | 2 | 252 | ||
AK040867.1 | 2 | 158 | BAC30725.1 | |
AK149935.1 | 3 | 297 | BAE29178.1 | |
AK150649.1 | 2 | 124 | BAE29736.1 | |
AK151482.1 | 2 | 124 | BAE30435.1 | |
AK152661.1 | 2 | 124 | BAE31397.1 | |
AK153304.1 | 2 | 158 | ||
AK155835.1 | 2 | 109 | ||
AK169494.1 | 2 | 158 | ||
AK171025.1 | 2 | 206 | BAE42194.1 | |
AK172222.1 | 2 | 100 | BAE42890.1 | |
BC035203.1 | 2 | 213 | AAH35203.2 | |
BY194349.1 | 2 | 205 | ||
CB248733.1 | 2 | 119 | ||
CF164288.1 | 2 | 196 |
Set Membership: |
Intragenic Exonic |