Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 18707 |
Gene Name: | phosphatidylinositol 3-kinase catalytic delta polypeptide |
Gene Aliases: |
2410099E07Rik, 2610208K16Rik, AW545373, p110delta |
Location: |
Chr.4:149649168-149702566 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pik3cd | NM_001029837.2 | NP_001025008.2 | ||
NM_001164049.1 | NP_001157521.1 | |||
NM_001164050.1 | NP_001157522.1 | |||
NM_001164051.1 | NP_001157523.1 | |||
NM_001164052.1 | NP_001157524.1 | |||
NM_008840.3 | NP_032866.2 | |||
AK040867.1 | BAC30725.1 | |||
AK149935.1 | BAE29178.1 | |||
AK153304.1 | ||||
AK171025.1 | BAE42194.1 | |||
AK172222.1 | BAE42890.1 | |||
BC035203.1 | AAH35203.2 | |||
CT010349.1 | CAJ18557.1 | |||
U86587.1 | AAC25676.1 |
Set Membership: |
Intragenic Non-exonic |