Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 18707 |
Gene Name: | phosphatidylinositol 3-kinase catalytic delta polypeptide |
Gene Aliases: |
2410099E07Rik, 2610208K16Rik, AW545373, p110delta |
Location: |
Chr.4:149649168-149702566 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pik3cd | NM_001029837.2 | 6 | 962 | NP_001025008.2 |
NM_001164049.1 | 6 | 989 | NP_001157521.1 | |
NM_001164050.1 | 6 | 989 | NP_001157522.1 | |
NM_001164051.1 | 6 | 1013 | NP_001157523.1 | |
NM_001164052.1 | 4 | 716 | NP_001157524.1 | |
NM_008840.3 | 7 | 1164 | NP_032866.2 | |
AK040867.1 | 6 | 924 | BAC30725.1 | |
AK149935.1 | 7 | 1063 | BAE29178.1 | |
AK150649.1 | 6 | 890 | BAE29736.1 | |
AK151482.1 | 6 | 890 | BAE30435.1 | |
AK152661.1 | 6 | 890 | BAE31397.1 | |
AK153304.1 | 6 | 924 | ||
AK155835.1 | 6 | 875 | ||
AK171025.1 | 6 | 972 | BAE42194.1 | |
AK172222.1 | 6 | 866 | BAE42890.1 | |
AK219741.1 | 2 | 106 | ||
BC035203.1 | 6 | 979 | AAH35203.2 | |
CT010349.1 | 4 | 684 | CAJ18557.1 | |
U86587.1 | 4 | 684 | AAC25676.1 |
Set Membership: |
Intragenic Exonic |