Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 209176 |
Gene Name: | indoleamine 2,3-dioxygenase 2 |
Gene Aliases: |
AI265623, C230043N17Rik, Ido-2, Indol1 |
Location: |
Chr.8:24531892-24576404 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ido2 | NM_145949.2 | NP_666061.3 | ||
AK082385.1 | ||||
BC026393.1 | AAH26393.1 | |||
EF137182.1 | ||||
EU440733.1 |
Set Membership: |
Intragenic Non-exonic |